Skip to Content
Merck
All Photos(17)

Key Documents

Safety Information

ZRB2350

Sigma-Aldrich

Anti-SCP-3 Antibody, clone 3M19 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Synaptonemal complex protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ICC
IF
IHC (p)
WB
affinity binding assay
clone:
3M19, recombinant monoclonal
species reactivity:
human
citations:
technique(s):
affinity binding assay: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3M19, recombinant monoclonal

description

3M19 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 27.73 kDa
observed mol wt ~25 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... SCP3(50511)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3M19 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Synaptonemal complex protein 3 (SCP-3). It targets an epitope within 25 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal region of human Synaptonemal complex protein 3 (SCP-3).

Application

Quality Control Testing

Evaluated by Western Blotting in Human testis tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SCP-3 in Human testis tissue lysate.

Tested Applications

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected SCP-3 in Human testis tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound SCP-3 peptide with a KD of 1.6 x 10-8 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected SCP-3 in Human testis tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SCP-3 in NTERA-2 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Synaptonemal complex protein 3 (UniProt: Q8IZU3; also known as SCP-3) is encoded by the SCP3 (also known as SYCP3) gene (Gene ID: 50511) in human. SCP-3 is a homotetrameric protein that is a component of the lateral elements of synaptonemal complex, a meiosis-specific protein structure essential for synapsis of homologous chromosomes. The tetrameric helix bundles assemble end to end into long homopolimeric fibers that exhibit a transversal striation with a periodicity of about 20 nm. It contains a central coiled-coil domain (aa 66-223) and a sequence in the C-terminal region (aa 231-236) that is important for its oligomerization and fiber formation. It is required for centromere pairing during meiosis in male germ cells and is essential for normal meiosis during spermatogenesis and male fertility. Male primordial germ cells are reported to express SCP-3 for a short time before undergoing G1 arrest. Overexpression of SCP-3 has been reported in various cancer cells and is associated with tumor stage and grade. Its overexpression promotes Akt-mediated tumorigenesis both in vivo and in vitro. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Cho, H., et al. (2014). PLoS ONE. 9(6); e98712; Di Carlo, AD., et al. (2000). Int. J. Dev. Biol. 44(2); 241-244).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

ZRB2350-25UL:
ZRB2350-4X25UL:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service