Merck
  • Evaluation of alcohol-based deep eutectic solvent in extraction and determination of flavonoids with response surface methodology optimization.
Evaluation of alcohol-based deep eutectic solvent in extraction and determination of flavonoids with response surface methodology optimization.
Journal of chromatography. A (2013-03-14)
Wentao Bi, Minglei Tian, Kyung Ho Row
ABSTRACT
Deep eutectic solvents (DESs) are emerging rapidly as a new type of green solvent instead of an ionic liquid (IL), and are typically formed by mixing choline chloride with hydrogen bond donors. Few studies have applied DESs to the extraction and determination of bioactive compounds. Therefore, in the present study, DESs were used to extract flavonoids (myricetin and amentoflavone), which are well known and widely used antioxidants, to extend their applications. A range of alcohol-based DESs with different alcohols to choline chloride (ChCl) mixing ratios were used for extraction using several extraction methods. Other factors, such as temperature, time, water addition and solid/liquid ratio, were examined systematically using a response surface methodology (RSM). A total of 0.031 and 0.518 mg g(-1) of myricetin and amentoflavone were extracted under the optimized conditions: 35 vol% of water in ChCl/1,4-butanediol (1/5) at 70.0 °C for 40.0 min and a solid/liquid ratio of 1/1 (g 10 mL(-1)). Good linearity was obtained from 0.1 × 10(-3) to 0.1 mg mL(-1) (r(2)>0.999). The excellent properties of DESs highlight their potential as promising green solvents for the extraction and determination of a range of bioactive compounds or drugs.
Product Description
Supelco
Amentoflavone, analytical standard
Supelco
Myricetin, analytical standard
Sigma-Aldrich
Amentoflavone, ≥98.0% (HPLC)
Sigma-Aldrich
1,4-Butanediol, ReagentPlus®, ≥99%
Sigma-Aldrich
1,4-Butanediol, ReagentPlus®, 99%
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)