Recommended Products
isotopic purity
98 atom % D
Quality Level
mp
112-114 °C (lit.)
mass shift
M+2
storage temp.
2-8°C
SMILES string
[2H]N([2H])c1cccc(c1)[N+]([O-])=O
InChI
1S/C6H6N2O2/c7-5-2-1-3-6(4-5)8(9)10/h1-4H,7H2/i/hD2
InChI key
XJCVRTZCHMZPBD-ZSJDYOACSA-N
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2 Oral
Target Organs
Kidney,Liver,Heart,Blood
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
384.8 °F - closed cup
Flash Point(C)
196.0 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service