NIST3132
Manganese (Mn) standard solution
NIST® SRM® 3132
Synonym(s):
Manganese (Mn) Standard Solution, Manganese nitrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
Related Categories
General description
This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of manganese. A unit of SRM 3132 consists of five 10 mL sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of manganese. The solution contains nitric acid at a volume fraction of approximately 10 %. For more information, please refer to the COA and SDS.
SRM 3132_cert SRM 3132 _SDS
SRM 3132_cert SRM 3132 _SDS
Other Notes
Certified for the analytes listed below.
Manganese (Mn)
See certificate for values and more details at nist.gov/SRM.
Manganese (Mn)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2
Supplementary Hazards
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service