SAB5700114
Anti-Acetyl-Histone H2B-K12 antibody produced in rabbit
Synonym(s):
GL105, H2B, H2B.1, H2BFQ, H2BGL105, H2BQ, HIST2H2BE, Histone H2B
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
13
species reactivity
human, mouse, rat
concentration
1.51 mg/ml
technique(s)
immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:2000
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
acetylation (Lys12)
Gene Information
human ... H2BC21(8349)
General description
Immunogen
Physical form
Storage and Stability
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service