Merck
Intermolecular clamping by hydrogen bonds: 2-pyridone⋅NH3.
Chemphyschem : a European journal of chemical physics and physical chemistry (2011-05-25)
Susan Blaser, Philipp Ottiger, Simon Lobsiger, Hans-Martin Frey, Samuel Leutwyler
ABSTRACT
A combined spectroscopic and ab initio theoretical study of the doubly hydrogen-bonded complex of 2-pyridone (2PY) with NH(3) has been performed. The S(1)←S(0) spectrum extends up to ≈1200 cm(-1) above the 0(0) (0) band, close to twice the range observed for 2PY. The S(1) state nonradiative decay for vibrations above ≈300 cm(-1) in the NH(3) complex is dramatically slowed down relative to bare 2PY. Also, the Δv=2, 4,… overtone bands of the ν(1) ' and ν(2) ' out-of-plane vibrations that dominate the low-energy spectral region of 2PY are much weaker or missing for 2PY⋅NH3, which implies that the bridging (2PY)NH⋅⋅⋅NH(3) and H(2) NH⋅⋅⋅O=C H-bonds clamp the 2PY at a planar geometry in the S(1) state. The mass-resolved UV vibronic spectra of jet-cooled 2PY⋅NH(3) and its H/D mixed isotopomers are measured using two-color resonant two-photon ionization spectroscopy. The S(0) and S(1) equilibrium structures and normal-mode frequencies are calculated by density functional (B3LYP) and correlated ab initio methods (MP2 and approximate second-order coupled-cluster, CC2). The S(1)←S(0) vibronic assignments are based on configuration interaction singles (CIS) and CC2 calculations. A doubly H-bonded bridged structure of C(S) symmetry is predicted, in agreement with that of Held and Pratt [J. Am. Chem. Soc. 1993, 115, 9718]. While the B3LYP and MP2 calculated rotational constants are in very good agreement with experiment, the calculated H(2) NH⋅⋅⋅O=C H-bond distance is ≈0.7 Å shorter than that derived by Held and Pratt. On the other hand, this underlines their observation that ammonia can act as a strong H-bond donor when built into an H-bonded bridge. The CC2 calculations predict the H(2) NH⋅⋅⋅O distance to increase by 0.2 Å upon S(1)←S(0) electronic excitation, while the (2PY)NH⋅⋅⋅NH(3) H-bond remains nearly unchanged. Thus, the expansion of the doubly H-bonded bridge in the excited state is asymmetric and almost wholly due to the weakening of the interaction of ammonia with the keto acceptor group.
Sigma-Aldrich
2-Hydroxypyridine, 97%
Levetiracetam impurity C, European Pharmacopoeia (EP) Reference Standard