Merck
  • Oligopeptide Transport in Rat Lung Alveolar Epithelial Cells is Mediated by Pept2.
Oligopeptide Transport in Rat Lung Alveolar Epithelial Cells is Mediated by Pept2.
Pharmaceutical research (2017-08-24)
Hovhannes J Gukasyan, Tomomi Uchiyama, Kwang-Jin Kim, Carsten Ehrhardt, Sharon K Wu, Zea Borok, Edward D Crandall, Vincent H L Lee
ABSTRACT
Studies were conducted in primary cultured rat alveolar epithelial cell monolayers to characterize peptide transporter expression and function. Freshly isolated rat lung alveolar epithelial cells were purified and cultured on permeable support with and without keratinocyte growth factor (KGF). Messenger RNA and protein expression of Pept1 and Pept2 in alveolar epithelial type I- and type II-like cell monolayers (±KGF, resp.) were examined by RT-PCR and Western blotting. RT-PCR showed expression of Pept2, but not Pept1, mRNA in both cell types. Western blot analysis revealed presence of Pept2 protein in type II-like cells, and less in type I-like cells. Bi-directional transmonolayer This study confirms the absence of Pept1 from rat lung alveolar epithelium in vitro. Functional Pept2 expression in type II-like cell monolayers suggests its involvement in oligopeptide lung disposition, and offers rationale for therapeutic development of di/tripeptides, peptidomimetics employing pulmonary drug delivery.
Sigma-Aldrich
Gly-Sar