Fluorescent Dye Wavelength Index
Fluorescent dyes are used to tag proteins, nucleic acids, and other biomolecules for use in life science applications, including fluorescence microscopy, flow cytometry, fluorescence in situ hybridization (FISH), receptor binding assays, and enzyme assays. Using the right combination of fluorescent-labeled biomolecules can enable multiple targets to be visualized simultaneously. Fluorescent dyes are optimized for excitation and detection in the visible light to near infrared spectrum. The choice of fluorochrome to use is determined by its excitation and emission wavelengths and the application.
To sort products within a column in the table below, click on the column heading. To re-alphabetize columns, refresh your web browser.
Product Name | Solvent | λ Ex Max | λ Em Max | Additional Conditions for Fluorescence | λ Ex Max | λ Em Max | |
---|---|---|---|---|---|---|---|
01673 | Acriflavine | EtOH | 463 | 490 | |||
06627 | 2-Aminoacridone | pH 8.0 | 420 | 538 | |||
08445 | 7-Amino-4-methyl-3-coumarinylacetic acid | MeOH | 350 | 433 | |||
08658 | 8-Aminonaphthalene-1,3,6-trisulfonic acid disodium salt | pH 7.0 | 356 | 512 | |||
09341 | 8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt | pH 7.4 | 420 | 500 | |||
25102 | 9-(Chloromethyl)anthracene | Carboxylic acid adducts | 365 | 412 | |||
10417 | 8-Anilino-1-naphthalenesulfonic acid ammonium salt | MeOH | 372 | 480 | Protein adduct | 372 | 480 |
10580 | Anthracene | CH3CN | 251 | 401 | |||
10609 | 9-Anthracenecarbonyl cyanide | MeOH | 361 | 460 | Hydroxyl compound adduct with ethanol | 361 | 451 |
75068 | 9,10-Anthracenediyl-bis(methylene)dimalonic acid | pH 7.0 | 380 | 407 | |||
89313 | Atto 390 | 390 | 475 | ||||
89204 | Atto 390 NHS ester | 389 | 471 | ||||
16805 | Atto 425-NHS ester | 437 | 483 | ||||
50712 | Atto 465 | ||||||
55607 | Atto 465 maleimide | 452 | 502 | ||||
53404 | Atto 465-NHS ester | 449 | 503 | ||||
41051 | Atto 488 | 504 | 521 | ||||
28562 | Atto 488 maleimide | 501 | 523 | ||||
41698 | Atto 488-NHS ester | 498 | 520 | ||||
16951 | Atto 495 | 499 | 535 | ||||
41022 | Atto 495 maleimide | 500 | 524 | ||||
00379 | Atto 495-NHS ester | 499 | 524 | ||||
77810 | Atto 520 NHS ester | H2O | 520 | 542 | EtOH | 525 | 547 |
06699 | Atto 532 | 533 | 560 | ||||
68499 | Atto 532 maleinimide | 532 | 553 | ||||
88793 | Atto 532 NHS ester | 530 | 551 | ||||
40592 | Atto 540 Q | ||||||
62453 | Atto 540 Q maleimide | ||||||
61683 | Atto 540 Q NHS ester | ||||||
42424 | Atto 550 | 554 | 574 | ||||
30730 | Atto 550 maleimide | 554 | 576 | ||||
92835 | Atto 550 NHS ester | 539 | 575 | ||||
75784 | Atto 565 | H2O | 561 | 585 | EtOH | 566 | 590 |
72464 | Atto 565 NHS ester | H2O | 561 | 585 | EtOH | 566 | 590 |
03722 | Atto 580 Q | ||||||
68152 | Atto 580 Q maleimide | ||||||
70425 | Atto 590 | H2O | 598 | 634 | EtOH | 598 | 634 |
79636 | Atto 590 NHS ester | H2O | 598 | 634 | EtOH | 598 | 634 |
08637 | Atto 594 | 601 | 627 | ||||
08717 | Atto 594 maleimide | 601 | 627 | ||||
08741 | Atto 594 NHS ester | 597 | 625 | ||||
78493 | Atto 610 | H2O | 605 | 646 | EtOH | 616 | 630 |
78493 | Atto 610 | 610 | 635 | ||||
41061 | Atto 610-maleimide | 610 | 637 | ||||
93259 | Atto 610 N-succinimidyl ester | H2O | 605 | 646 | EtOH | 616 | 630 |
93259 | Atto 610 N-succinimidyl ester | 610 | 635 | ||||
04327 | Atto 612 Q | ||||||
50332 | Atto 612 Q maleimide | ||||||
53988 | Atto 612 Q NHS ester | ||||||
92716 | Atto 620 | 617 | 641 | ||||
67351 | Atto 620 NHS ester | 620 | 641 | ||||
18620 | Atto 633 | ||||||
01464 | Atto 633 NHS ester | 630 | 650 | ||||
08968 | Atto 635 | 635 | 656 | ||||
97875 | Atto 647 | 643 | 665 | ||||
41784 | Atto 647 maleimide | 647 | 669 | ||||
04507 | Atto 647N | 644 | 667 | ||||
07376 | Atto 647 NHS ester | 643 | 662 | ||||
05316 | Atto 647N maleimide | 644 | 669 | ||||
18373 | Atto 647N NHS ester | 647 | 661 | ||||
93711 | Atto 655 | H2O | 665 | 690 | EtOH | 655 | 680 |
76245 | Atto 655 NHS ester | H2O | 665 | 690 | EtOH | 655 | 680 |
75999 | Atto 680 NHS ester | H2O | 680 | 702 | EtOH | 675 | 699 |
30674 | Atto 700 | 681 | 714 | ||||
50611 | Atto 700 maleimide | 645 | 715 | ||||
16986 | Atto 700 NHS-ester | 681 | 713 | ||||
47156 | Atto 725 | 725 | 752 | ||||
93725 | Atto 725 NHS ester | 725 | 753 | ||||
91394 | Atto 740 | 740 | 764 | ||||
14448 | 2-(4-Biphenylyl)-6-phenylbenzoxazole | H2O | 425 | CH3CN, MeOH | 390 | ||
14510 | 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid | pH 8.0 | 287 | 363 | with calcium | 274 | 363 |
14513 | 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetrasodium salt | pH 8.0 | 254 | 363 | with calcium | 274 | 363 |
14530 | bisBenzimide H 33258 | pH 7.0 | 338 | 505 | bound to DNA | 356 | 465 |
14533 | bisBenzimide H 33342 trihydrochloride | pH 7.0 | 340 | 510 | bound to DNA | 355 | 465 |
14562 | 2',7'-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein tetrakis(acetoxymethyl) ester | Reaction product | 482 | 528 | |||
15025 | meso-1,2-Bis(4-methoxyphenyl)ethylenediamine | Saccharide adducts | 340 | 460 | |||
15090 | 1,4-Bis(2-methylstyryl)benzene | Cyclohexane | 419 | Heptane | 416 | ||
69898 | Bromobimane | pH 7.0 | 398 | Thiol adduct | 390 | 478 | |
72301 | 9-(2-Carboxy-2-cyanovinyl)julolidine | pH 8.0 | 433 | 500 | |||
21882 | 5(6)-Carboxy-2',7'-dichlorofluorescein | pH 8.0 | 504 | 529 | |||
21884 | 5(6)-Carboxy-2',7'-dichlorofluorescein diacetate | Reaction product | 470 | 504 | |||
86826 | 5-Carboxyfluorescein | pH 8.0 | 492 | 517 | |||
21877 | 5(6)-Carboxyfluorescein | pH > 8.0 | 490 | 520 | |||
21879 | 5(6)-Carboxyfluorescein diacetate | hydrolyzed | 492 | 517 | |||
21888 | 5(6)-Carboxyfluorescein diacetate N-succinimidyl ester | DMF | <300 | hydrolyzed | 492 | 517 | |
21878 | 5(6)-Carboxyfluorescein N-hydroxysuccinimide ester | pH 9.0 | 495 | 519 | |||
92846 | 5-Carboxyfluorescein N-succinimidyl ester | pH 8.0 | 492 | 517 | |||
21965 | 5(6)-Carboxy-X-rhodamine | MeOH | 568 | 595 | |||
41969 | 5-Carboxy-X-rhodamine N-succinimidyl ester | pH 7.0 | 575 | 600 | |||
21953 | 5(6)-Carboxytetramethylrhodamine | MeOH | 541 | 566 | |||
55222 | 6-Carboxytetramethylrhodamine | pH 8.0 | 543 | 575 | |||
53048 | 5-Carboxy-tetramethylrhodamine N-succinimidyl ester | pH 7.0 | 543 | 575 | |||
21955 | 5(6)-Carboxytetramethylrhodamine N-succinimidyl ester | MeOH | 540 | 565 | |||
88997 | 6-Carboxy-tetramethylrhodamine N-succinimidyl ester | pH 7.0 | 543 | 575 | |||
25455 | 4-Chloro-7-nitrobenzofurazan | MeOH | 336 | Aliphatic amine adducts | 475 | 540 | |
30423 | 3-(Dansylamino)phenylboronic acid | MeOH | 337 | 517 | Protein adduct | 337 | 490 |
30432 | Dansylcadaverine | MeOH | 335 | 518 | Reaction product | 338 | 500 |
39220 | Dansyl chloride | CHCl3 | 372 | Butylamine adduct | 337 | 492 | |
30434 | Dansylhydrazine | MeOH | 336 | 534 | Saccharide adducts | 336 | 487 |
32670 | 4',6-Diamidino-2-phenylindole dihydrochloride | H2O | bound to DNA | 358 | 461 | ||
34025 | Dibromobimane | MeOH | 391 | Thiol adduct | 391 | 480 | |
72335 | 9-(2,2-Dicyanovinyl)julolidine | pH 8.0 | 433 | 500 | |||
36798 | 7-(Diethylamino)coumarin-3-carbohydrazide | MeOH | 420 | 468 | Carboxylic acid adduct | 415 | 480 |
31755 | 7-(Diethylamino)coumarin-3-carbonyl azide | MeOH | 432 | 480 | |||
36799 | 7-(Diethylamino)coumarin-3-carboxylic acid | pH 9.0 | 409 | 473 | |||
36801 | 7-(Diethylamino)coumarin-3-carboxylic acid N-succinimidyl ester | MeOH | 432 | 472 | |||
37150 | 3,3'-Diheptylthiacarbocyanine iodide | MeOH | 562 | 575 | |||
37291 | Dihydroethidium | MeCN | 355 | 420 | bound to DNA | 358 | 461 |
37920 | 6,8-Dihydroxy-1,3-pyrenedisulfonic acid disodium salt | pH 4.0 | 405 | 465 | pH 8.0 | 458 | 500 |
39235 | 11-[5-(Dimethylamino)-1-naphthalenesulfonylamino]undecanoic acid | MeOH | 335 | 519 | |||
39255 | 4-Dimethylamino-4'-nitrostilbene | CH2Cl2 | 500 | 700 | |||
41525 | N,N-Dimethyl-6-propionyl-2-naphthylamine | MeOH | 361 | 498 | |||
41785 | Dimidium bromide | pH 7.6, low ionic strength | 480 | Nucleic acid adducts, Tris pH 8.0 | 306 | 612 | |
42364 | 1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate | MeOH | 550 | 565 | |||
43608 | 3,3'-Dipropylthiadicarbocyanine iodide | MeOH | 651 | 675 | |||
40227 | 6-Dodecanoyl-N,N-dimethyl-2-naphthylamine | MeOH | 364 | 497 | |||
44583 | Doxorubicin hydrochloride | EtOH | 470 | 585 | |||
45245 | Eosin 5-isothiocyanate | pH 9.0 | 521 | 544 | |||
46067 | Ethidium bromide solution | H2O | 480 | 620 | bound to DNA | 518 | 605 |
46043 | Ethidium homodimer | H2O | 493 | bound to DNA | 528 | 617 | |
46123 | 1-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide | H2O | 355 | 460 | |||
46125 | 8-Ethoxy-N,N,N',N',N'',N''-hexamethylpyrene-1,3,6-trisulfonamide | Reaction product | 380 | 520 | |||
46393 | Fluo-3 | pH > 6.0 | 503 | with calcium | 506 | 526 | |
46917 | 9-Fluorenylmethyl carbazate | MeOH | 264 | 309 | Saccharide adducts | 270 | 320 |
46955 | Fluorescein | pH 9.0 | 490 | 514 | |||
46960 | Fluorescein sodium salt | pH 9.0 | 490 | 514 | |||
46935 | 6-[Fluorescein-5(6)-carboxamido]hexanoic acid | pH 9.0 | 480 | 525 | |||
46940 | 6-[Fluorescein-5(6)-carboxamido]hexanoic acid N-hydroxysuccinimide ester | pH 8.2 | 491 | 515 | |||
46943 | Fluorescein dilaurate | Reaction product | 490 | 514 | |||
46950 | Fluorescein 5(6)-isothiocyanate | pH 9.0 | 495 | 520 | |||
46946 | Fluorescein isothiocyanate–dextran 150000 | pH 8.0 | 490 | 520 | |||
46945 | Fluorescein isothiocyanate–dextran 70000 | pH 8.0 | 490 | 520 | |||
46947 | Fluorescein isothiocyanate – Dextran 500000-Conjugate | pH 8.0 | 490 | 520 | |||
46985 | Fluorescein-5-thiosemicarbazide | pH 9.0 | 492 | 516 | |||
55817 | Fluorescent red 630 | pH 7.0 | 620 | 650 | |||
47140 | 4-Fluoro-7-nitrobenzofurazan | EtOH | 470 | 528 | Amino acid adducts | 470 | 530 |
47614 | Fluram® | pH 9.0 | 234 | Protein adduct | 334 | 455 | |
52335 | N-Hexadecylpyrene-1-sulfonamide | MeOH | 350 | 400 | |||
55155 | 7-Hydroxycoumarin-3-carboxylic acid | pH 4.0 | 342 | 447 | pH 9.0 | 386 | 448 |
55156 | 7-Hydroxycoumarin-3-carboxylic acid N-succinimidyl ester | MeOH | 418 | 447 | |||
55634 | 7-Hydroxy-4-methyl-3-coumarinylacetic acid | pH 10.0 | 360 | 445 | |||
55627 | 7-Hydroxy-4-methyl-2(1H)-quinolone | pH 5.0 | 321 | 357 | pH 9.0 | 351 | 438 |
56059 | 7-Hydroxy-N-octadecylcoumarin-3-carboxamide | CHCl3/DMSO | 359 | 406 | |||
57180 | Indo 1 | 100 mM KCl | 349 | 482 | with calcium | 331 | 398 |
64951 | 7-Methoxycoumarin | CHCl3 | 340 | 389 | Reaction product | 360 | 449 |
64948 | 7-Methoxycoumarin-3-carboxylic acid | MeOH | 360 | 415 | |||
64958 | 2-Methoxy-2,4-diphenyl-3(2H)-furanone | Amine adducts | 390 | 480 | |||
69025 | 1-Methylpyrene | CHCl3 | 340 | 486 | |||
69587 | 2'-(4-Methylumbelliferyl)-a- D -N-acetylneuraminic acid sodium salt hydrate | pH 7.0 | 315 | 374 | Reaction product | 360 | 449 |
69585 | 4-Methylumbelliferyl N-acetyl-ß- D -glucosaminide dihydrate | pH 10.0 | 317 | Reaction product | 360 | 449 | |
19362 | 4-Methylumbelliferyl butyrate | MeOH | 312 | Reaction product | 360 | 449 | |
69591 | 4-Methylumbelliferyl a- D -glucopyranoside | pH 9.0 | 317 | 374 | Reaction product | 360 | 449 |
69602 | 4-Methylumbelliferyl-ß- D -glucuronide hydrate | pH 9.0 | 316 | 375 | Reaction product | 360 | 449 |
51010 | 4-Methylumbelliferyl 4-guanidinobenzoate hydrochloride hydrate | pH 6.0 | 323 | 464 | Reaction product | 360 | 449 |
75164 | 4-Methylumbelliferyl oleate | MeOH | 312 | Reaction product | 360 | 449 | |
69899 | Monochlorobimane | MeOH | 380 | 461 | Thiol adduct | 380 | 480 |
70420 | Naphthofluorescein | pH 10.0 | 594 | 663 | |||
70450 | 2-Naphthol | 0.1 M NaOH | 320 | 410 | |||
70448 | 2-Naphthol | 0.1 M NaOH | 320 | 410 | |||
70684 | 1-(2-Naphthoyl)imidazole | H2O | 230 | 362 | Aldite adducts | 234 | 374 |
15167 | NIR-797 isothiocyanate | MeOH | 668 | 798 | |||
72963 | 12-(7-Nitrobenzofurazan-4-ylamino)dodecanoic acid | MeOH | 467 | 539 | |||
72964 | 6-(7-Nitrobenzofurazan-4-ylamino)hexanoic acid | MeOH | 467 | 539 | |||
74758 | 8-Octadecyloxypyrene-1,3,6-trisulfonic acid trisodium salt | H2O | 404 | 434 | |||
74875 | 8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt | H2O | 384 | 409 | Reaction product | 460 | 520 |
75926 | Oxonol VI | MeOH | 599 | 634 | |||
76275 | PBFI-AM | MeOH | 369 | with Na | 334 | 525 | |
77490 | Phenanthridine | DMSO | 265 | 415 | |||
79279 | 3-Phenylumbelliferone | pH 3.0 | 338 | 463 | pH 10.0 | 383 | 472 |
79760 | Phthaldialdehyde | pH 7.0 | 257 | Thiol adduct | 334 | 455 | |
81845 | Propidium iodide | pH 7.2 | 482 | 608 | bound to DNA | 540 | 608 |
82648 | Pyrene | n-Pentane | 350 | 392 | |||
82660 | 1-Pyrenedecanoic acid | MeOH | 341 | 377 | |||
82663 | 1-Pyrenedodecanoic acid | MeOH | 341 | 377 | |||
82657 | 1-Pyrenesulfonic acid sodium salt | MeOH | 344 | 376 | |||
08520 | Quin-2 | with calcium | 332 | 493 | |||
22620 | Quinine | 0.5 M Sulfuric acid | 347 | 448 | |||
22640 | Quinine hemisulfate salt monohydrate | H2O | 343 | 453 | |||
22630 | Quinine hydrochloride dihydrate | H2O | 322 | 450 | |||
83636 | Resorufin acetate | DMSO | 540 | 590 | Reaction product | 571 | 585 |
83637 | Resorufin butyrate | DMSO | 540 | 590 | Reaction product | 571 | 585 |
46121 | Resorufin ethyl ether | MeOH | 464 | ||||
69125 | Resorufin methyl ether | MeOH | 463 | ||||
83689 | Rhodamine B | Acidic EtOH | 554 | 627 | |||
83697 | Rhodamine 6G | MeOH | 528 | 551 | |||
83694 | Rhodamine 101 | MeOH | 565 | 589 | |||
83695 | Rhodamine 110 chloride | MeOH | 499 | 521 | |||
83701 | Rhodamine 800 | MeOH | 528 | 547 | |||
83685 | Rhodamine B octadecyl ester perchlorate | MeOH | 554 | 578 | |||
17866 | Streptavidin-CryptoFluor™ Crimson | pH 7.2 | 585 | 658 | |||
86186 | Sulforhodamine B acid chloride | H2O | 565 | 586 | |||
86470 | p-Terphenyl | EtOH | 277 | ||||
87917 | Tetramethylrhodamine ethyl ester perchlorate | MeOH | 549 | 574 | |||
87918 | Tetramethylrhodamine isothiocyanate mixed isomers | MeOH | 543 | 571 | |||
88071 | meso-Tetraphenylporphyrin | Toluene | 645 | 651 | |||
43060 | N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium p-toluenesulfonate | MeOH | 355 | 450 | |||
93979 | Umbelliferone | 95% EtOH | 325 | 441 |