Skip to Content
Merck
All Photos(1)

Key Documents

82660

Sigma-Aldrich

1-Pyrenedecanoic acid

suitable for fluorescence, ≥98.0% (HPLC)

Synonym(s):

10-(1-Pyrenyl)decanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28O2
CAS Number:
Molecular Weight:
372.50
Beilstein:
2007670
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

≥98.0% (HPLC)

form

powder

fluorescence

λex 340 nm; λem 376 nm in methanol

suitability

suitable for fluorescence

SMILES string

OC(=O)CCCCCCCCCc1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C26H28O2/c27-24(28)12-7-5-3-1-2-4-6-9-19-13-14-22-16-15-20-10-8-11-21-17-18-23(19)26(22)25(20)21/h8,10-11,13-18H,1-7,9,12H2,(H,27,28)

InChI key

PIMKEUIWMFJVNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

In addition to the emission maximum at 376 nm, there are lower maxima at 396, 415 and 440 nm.

Other Notes

Anionic membrane probe[1]; Measurement of membrane fluidity of polymorphonuclear leukocytes by flow cytometry[2]; Biosynthetically incorporated into cultured fibroblasts[3]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Masuda et al.
Journal of immunological methods, 96(2), 225-231 (1987-02-11)
A method was established for measuring membrane fluidity of polymorphonuclear leukocytes (PMN) by the excimer-forming lipid technique with pyrenedecanoic acid in flow cytometry. When cells were labeled, the use of 2-25 microM of pyrenedecanoic acid provided similar results. Neither the
J Radom et al.
Biochimica et biophysica acta, 920(2), 131-139 (1987-07-31)
The lipid metabolism in cultured fibroblasts from multisystemic (type 3) lipid storage myopathy and controls has been studied through pulse-chase experiments using 1-pyrenedecanoic acid as precursor. The uptake of 1-pyrenedecanoic acid was not significantly different in multisystemic lipid storage myopathy
Pyrenedecanoic acid and pyrene lecithin.
H J Galla et al.
Methods in enzymology, 72, 471-479 (1981-01-01)
Amit K Saha et al.
Journal of biomechanics, 52, 83-88 (2017-01-14)
The role of cholesterol content on monocyte biomechanics remains understudied despite the well-established link between cholesterol and monocytes/macrophages in atherosclerosis, and the effect on other cell types. In this work, we have investigated the effect of cholesterol on monocyte deformability
Taro Yasumoto et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(8), 9220-9234 (2019-05-16)
Amyloid β-protein (Aβ) molecules tend to aggregate and subsequently form low MW (LMW) oligomers, high MW (HMW) aggregates such as protofibrils, and ultimately fibrils. These Aβ species can generally form amyloid plaques implicated in the neurodegeneration of Alzheimer disease (AD)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service