Skip to Content
MilliporeSigma

AABH97CD70B7

AA BLOCKS, INC.

Benzoic acid, 2-hydroxy-, compd. with (3aS,8aR)-1,2,3,3a,8,8a-hexahydro-1,3a,8-trimethylpyrrolo[2,3-b]indol-5-yl N-methylcarbamate (1:1)

Synonym(s):

(3aS)-1,3a,8-trimethyl-1,2,3,3a,8,8a-hexahydropyrrolo[2,3-b]indol-5-yl methylcarbamate 2-hydroxybenzoate

Select a Size

Change View

Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H21N3O2.C7H6O3/c1-15-7-8-17(3)13(15)18(4)12-6-5-10(9-11(12)15)20-14(19)16-2;8-6-4-2-1-3-5(6)7(9)10/h5-6,9,13H,7-8H2,1-4H3,(H,16,19);1-4,8H,(H,9,10)/t13?,15-;/m0./s1

Inchi Key

HZOTZTANVBDFOF-LDCKTULKSA-N

Purity

98%

Storage temp.

2~8C

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library