Skip to Content
MilliporeSigma

AABH97CD8588

AA BLOCKS, INC.

FR900098 (sodium salt)

Synonym(s):

sodium hydrogen 3-[acetyl(hydroxy)amino]propylphosphonate

Select a Size

Change View

About This Item

Linear Formula:
C5H11O5N1P1Na1
CAS Number:
Molecular Weight:
219.11
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H12NO5P.Na/c1-5(7)6(8)3-2-4-12(9,10)11;/h8H,2-4H2,1H3,(H2,9,10,11);/q;+1/p-1

Inchi Key

LCFXFDGYDKNWMD-UHFFFAOYSA-M

Purity

95%

Country of Origin

US

Storage temp.

-20C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library