Skip to Content
MilliporeSigma

ATEH03AB2B98

AstaTech, Inc.

SODIUM OLEATE

Synonym(s):

sodium (9E)-9-octadecenoate

Select a Size

Change View

About This Item

Linear Formula:
C18H33O2Na1
CAS Number:
Molecular Weight:
304.45
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Country of Origin

CN

Shipping Temperature

Normal

Purity

95%

Storage temp.

2-8 C

Physical Form

White to Yellow Solid

Inchi Code

1S/C18H34O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h9-10H,2-8,11-17H2,1H3,(H,19,20);/q;+1/p-1/b10-9+;

Inchi Key

BCKXLBQYZLBQEK-RRABGKBLSA-M



Still not finding the right product?

Explore all of our products under SODIUM OLEATE


Precautionary Statements

Signal Word

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library