Skip to Content
MilliporeSigma

ATEH11AB24BB

AstaTech, Inc.

1,7-OCTADIENE

Synonym(s):

1,7-octadiene

Select a Size

Change View

About This Item

Linear Formula:
C8H14
CAS Number:
Molecular Weight:
110.2
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H14/c1-3-5-7-8-6-4-2/h3-4H,1-2,5-8H2

Inchi Key

XWJBRBSPAODJER-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

2-8 C

Country of Origin

CN



Still not finding the right product?

Explore all of our products under 1,7-OCTADIENE


Precautionary Statements

Signal Word

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library