Inchi Code
1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1
Inchi Key
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
2-8 C
Country of Origin
US
Physical Form
Solid
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.