Inchi Code
1S/C13H20O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h7-8H,5-6,9H2,1-4H3/b8-7+
Inchi Key
PSQYTAPXSHCGMF-BQYQJAHWSA-N
Shipping Temperature
Normal
Purity
95%
Country of Origin
CN
Physical Form
Liquid
Storage temp.
Sealed in dry, 2-8C
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
