Skip to Content
MilliporeSigma

BL3H1F1C9906

BLD Pharmatech

Imidazo[1,2-a]pyridine

Synonym(s):

imidazo[1,2-a]pyridine

Select a Size

Change View

About This Item

Linear Formula:
C7H6N2
CAS Number:
Molecular Weight:
118.14
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H6N2/c1-2-5-9-6-4-8-7(9)3-1/h1-6H

Inchi Key

UTCSSFWDNNEEBH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: liquid / Colour: Pale yellow - Deep yellow green - Brown - Black



Still not finding the right product?

Explore all of our products under Imidazo[1,2-a]pyridine


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library