Skip to Content
MilliporeSigma

BL3H1F1C9F07

BLD Pharmatech

EDTA disodium salt

Synonym(s):

sodium 2,2'-(2-(bis(carboxymethyl)amino)ethylazanediyl)diacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H14O8N2Na2
CAS Number:
Molecular Weight:
336.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H16N2O8.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;2*+1/p-2

Inchi Key

ZGTMUACCHSMWAC-UHFFFAOYSA-L

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White powder



Still not finding the right product?

Explore all of our products under EDTA disodium salt



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library