Skip to Content
MilliporeSigma

BL3H1F1CE04B

BLD Pharmatech

5,5'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(isobenzofuran-1,3-dione)

Synonym(s):

5-[4-(1-{4-[(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)oxy]phenyl}-1-methylethyl)phenoxy]-2-benzofuran-1,3-dione

Select a Size

Change View

About This Item

Linear Formula:
C31H20O8
CAS Number:
Molecular Weight:
520.5
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C31H20O8/c1-31(2,17-3-7-19(8-4-17)36-21-11-13-23-25(15-21)29(34)38-27(23)32)18-5-9-20(10-6-18)37-22-12-14-24-26(16-22)30(35)39-28(24)33/h3-16H,1-2H3

Inchi Key

MQAHXEQUBNDFGI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder

Melting Point

184-187C



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library