Skip to Content
MilliporeSigma

BL3H953890F4

BLD Pharmatech

4-Amino-1-benzylpiperidine

Synonym(s):

1-benzyl-4-piperidinamine

Select a Size

Change View

About This Item

Linear Formula:
C12H18N2
CAS Number:
Molecular Weight:
190.29
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H18N2/c13-12-6-8-14(9-7-12)10-11-4-2-1-3-5-11/h1-5,12H,6-10,13H2

Inchi Key

YUBDLZGUSSWQSS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 4-Amino-1-benzylpiperidine


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library