Inchi Code
1S/2C2H4O2.Cd.2H2O/c2*1-2(3)4;;;/h2*1H3,(H,3,4);;2*1H2/q;;+2;;/p-2
Inchi Key
AUIZLSZEDUYGDE-UHFFFAOYSA-L
Shipping Temperature
Normal
Purity
99%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Form: solid
Still not finding the right product?
Explore all of our products under Cadmium acetate dihydrate
hstatements
Precautionary Statements
Signal Word
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


