Skip to Content
MilliporeSigma

BL3H97A47718

BLD Pharmatech

Nonan-5-one

Synonym(s):

5-nonanone

Select a Size

Change View

About This Item

Linear Formula:
C9H18O1
CAS Number:
Molecular Weight:
142.24
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H18O/c1-3-5-7-9(10)8-6-4-2/h3-8H2,1-2H3

Inchi Key

WSGCRAOTEDLMFQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: liquid / Colour: Colorless - Pale yellow



Still not finding the right product?

Explore all of our products under Nonan-5-one




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library