Skip to Content
MilliporeSigma

BL3H9AA51DF6

BLD Pharmatech

Polycaprolactone

Synonym(s):

2-oxepanone

Select a Size

Change View

About This Item

Linear Formula:
C6H10O2
CAS Number:
Molecular Weight:
114.14
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H10O2/c7-6-4-2-1-3-5-8-6/h1-5H2

Inchi Key

PAPBSGBWRJIAAV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

80000

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White beads



Still not finding the right product?

Explore all of our products under Polycaprolactone


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library