Skip to Content
MilliporeSigma

COM448677511

Combi-Blocks, Inc.

Guanosine 5'-diphosphate mono sodium salt

Synonym(s):

sodium ((2R,3S,4R,5R)-5-(2-amino-6-hydroxy-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogendiphosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
465.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H15N5O11P2.Na/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(25-9)1-24-28(22,23)26-27(19,20)21;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H2,19,20,21)(H3,11,13,14,18);/q;+1/p-1/t3-,5-,6-,9-;/m1./s1

Inchi Key

AGZYQLYGEWDYOF-GWTDSMLYSA-M

Storage temp.

Refrigerated

Purity

95%

Country of Origin

CN



MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?SS-8623



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library