Skip to Content
MilliporeSigma

ZRB3113

Anti-Myostatin Antibody, clone 1F10 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells

Sustainability features

enhanced validation

Enhanced Validation

Synonym(s):

GDF-8, Growth/differentiation factor 8

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1F10, recombinant monoclonal
Application:
ELISA, IHC, WB, affinity binding assay
Citations:
-
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F10, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 42.75 kDa, observed mol wt ~14 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

porcine, horse, feline, rat, mouse, monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Sustainability features

technique(s)

ELISA: suitable, affinity binding assay: suitable, immunohistochemistry: suitable, western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... MSTN(2660)

General description

Growth/differentiation factor 8 (UniProt: O14793; also known as GDF-8, Myostatin) is encoded by the MSTN (also known as GDF8) gene (Gene ID: 2660) in human. Myostatin is a secreted homodimeric glycoprotein of the transforming growth factor β (TGF β) superfamily that functions as a key negative regulator of skeletal muscle mass and broader musculoskeletal homeostasis. It is synthesized primarily in skeletal muscle as a pre propeptide comprising an N-terminal signal peptide, a large pro domain, and a C terminal growth factor domain. After signal peptide removal and furin like protease cleavage, the C-terminal dimer remains non covalently bound to the pro domain as a latent complex, which can be activated extracellularly by metalloproteinases and other proteases to release mature myostatin. The active C-terminal dimer adopts the canonical TGF β like "cysteine knot" fold and signals by binding activin type II receptors (ActRIIA/ACVR2A or ActRIIB/ACVR2B), which then recruit type I receptors ALK4 or ALK5 to form a heterotetrameric complex that phosphorylates receptor regulated SMAD2/3. These associate with SMAD4 and translocate to the nucleus to repress anabolic programs (e.g. IGF1/AKT–mTOR signaling) and induce catabolic and cell cycle inhibitory genes such as CDKN1A (p21). Myostatin expression is highest in skeletal muscle but is also detectable in cardiac muscle, adipose tissue, and several non muscle organs. It circulates in serum predominantly in latent complexes, and acts in autocrine, paracrine and endocrine fashions. In myogenic progenitors and myoblasts, myostatin suppresses proliferation and differentiation by activating SMAD2/3 and, secondarily, p38 MAPK and PI3K–AKT/mTOR pathways, thereby maintaining quiescence and limiting myofiber hypertrophy. Genetic deletion or pharmacological blockade of GDF8 in mice, livestock and humans leads to dramatic skeletal muscle hypertrophy and hyperplasia, establishing myostatin as a dominant physiological brake on muscle growth; myostatin also skews mesenchymal progenitors toward adipogenesis and away from myogenesis, linking it to sarcopenic and cachectic body composition changes. Beyond muscle, Myostatin exerts direct effects on bone and cartilage. It inhibits osteogenic differentiation, suppresses chondrocyte proliferation, and promotes osteoclastogenesis by engaging ActRII receptors on osteoclast precursors and driving a SMAD2/3–NFATc1 program, thereby enhancing RANKL dependent osteoclast differentiation and bone resorption. Myostatin deficiency or inhibition reduces inflammatory joint destruction in murine models of rheumatoid arthritis and mitigates post traumatic osteoarthritis after ACL injury, indicating that it is an important mediator of muscle–bone–joint crosstalk in musculoskeletal degeneration. Pathologically, serum and muscle myostatin levels are elevated in chronic kidney disease, cancer, sepsis, AIDS and other chronic inflammatory states, where they correlate with muscle wasting and functional decline and likely contribute causally to cachexia and sarcopenia. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. 
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.
This product aligns with one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals, and exceptionally stable to allow for ambient shipping and storage, and thus aligns with Green Chemistry Principles ′Waste Prevention′, ′Designing Safer Chemicals′, and ′Design for Energy Efficiency′.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal half of human Myostatin.

Application

Quality Control Testing
Evaluated by Western Blotting with a recombinant fragment of human Myostatin.
Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected a recombinant fragment of human Myostatin.
Tested Applications
Western Blotting Analysis: A 1:100 dilution of this antibody detected Myostatin in human heart tissue lysate.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Myostatin in human skeletal muscle tissue sections.
Enzyme Immunoassay: Various of dilutions of a representative lot of this antibody detected Myostatin protein in ELISA application.
Affinity Binding Assay: A representative lot of this antibody bound with Myostatin peptide with at least one thousand-fold (1,000X) higher affinity than with a non-specific control peptide.
Note: Actual optimal working dilutions must be determined by end user as specimens and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 1F10 is a ZooMAb® rabbit recombinant monoclonal antibody that detects Myostatin. It targets an epitope within 17 amino acids from the C-terminal half.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library