Select a Size
About This Item
biological source
rabbit
Quality Segment
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
5G17, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 17.18 kDa, observed mol wt ~17 kDa
purified by
using Protein A
species reactivity
human
species reactivity (predicted by homology)
boar, nonhuman primates
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Sustainability features
technique(s)
affinity binding assay: suitable, immunofluorescence: suitable, immunohistochemistry: suitable, western blot: suitable
isotype
IgG
epitope sequence
N-terminal half
Protein ID accession no.
UniProt accession no.
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... MB(4151)
General description
Immunogen
Application
Evaluated by Western Blotting in human skeletal muscle tissue lysate.
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Myoglobin in human skeletal muscle tissue lysate.
Tested Applications
Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected Myoglobin in human heart tissue lysate.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Myoglobin in human skeletal muscle tissue sections.
Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected Myoglobin in human skeletal muscle tissue sections.
Affinity Binding Assay: A representative lot of this antibody bound with Myoglobin peptide with at least one thousand-fold (1,000X) higher affinity than with a non-specific control peptide.
Note: Actual optimal working dilutions must be determined by end user as specimens and experimental conditions may vary with the end user.
Biochem/physiol Actions
Physical form
Preparation Note
Legal Information
Disclaimer
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.