Skip to Content
Merck
All Photos(1)

Key Documents

1.05418

Millipore

Bismuth sulfite agar acc. to WILSON-BLAIR

for the isolation and differentiation of Salmonella typhi

Synonym(s):

Bismuth-sulfite agar acc. to WILSON-BLAIR, WILSON-BLAIR medium

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41106213

Quality Level

form

powder (dehydrated (DCM))

technique(s)

microbiological culture: suitable

pH

7.6 (37 °C, 47 g/L in H2O, after autoclaving)

solubility

47 g/L

bulk density

680 kg/m3

application(s)

clinical
microbiology

storage temp.

15-25°C

suitability

Salmonella spp.

General description

for the isolation and differentiation of Salmonella typhi

Analysis Note

Appearance (clearness)turbidAppearance (colour)greenSolidification behaviour (2 hrs., 45 °C)liquidpH-value (25 °C)7.4 - 7.8Growth (Salmonella typhimurium ATCC 14028 (WDCM 00031))good to very goodGrowth (Salmonella typhimurium ATCC 13311 (WDCM 00121))good to very goodGrowth (Salmonella Abony NCTC 6017)good to very goodGrowth (Salmonella enteritidis NCTC 5188 (WDCM 00119))good to very goodGrowth (Salmonella arizonae ATCC 13314)fair to very goodGrowth (Escherichia coli ATCC 25922 (WDCM 00013))poor to fairGrowth (Proteus mirabilis ATCC 29906 (WDCM 00023))good to very goodGrowth (Shigella sonnei ATCC 11060)noneGrowth (Staphylococcus aureus ATCC 25923 (WDCM 00034))noneGrowth (Bacillus cereus ATCC 11778 (WDCM 00001))noneColonies (Salmonella typhimurium ATCC 14028 (WDCM 00031))blackColonies (Salmonella typhimurium ATCC 13311 (WDCM 00121))blackColonies (Salmonella Abony NCTC 6017)blackColonies (Salmonella enteritidis NCTC 5188 (WDCM 00119))blackColonies (Salmonella arizonae ATCC 13314)blackColonies (Escherichia coli ATCC 25922 (WDCM 00013))greenColonies (Proteus mirabilis ATCC 29906 (WDCM 00023))oliveMetallic lustre (Salmonella typhimurium ATCC 14028 (WDCM 00031))+Metallic lustre (Salmonella typhimurium ATCC 13311 (WDCM 00121))+Metallic lustre (Salmonella Abony NCTC 6017)+Metallic lustre (Salmonella enteritidis NCTC 5188 (WDCM 00119))+Metallic lustre (Salmonella arizonae ATCC 13314)+Incubation: 48 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service