59808
Atto 740 NHS ester
BioReagent, suitable for fluorescence, ≥90.0% (coupling to amines, HPLC)
About This Item
Recommended Products
product line
BioReagent
Assay
≥90.0% (coupling to amines, HPLC)
manufacturer/tradename
ATTO-TEC GmbH
transmittance
254 nm, >90%
740 nm, >90%
fluorescence
λex 740 nm; λem 766 nm in 0.1 M phosphate pH 7.0
λ
in ethanol (with 0.1% trifluoroacetic acid)
UV absorption
λ: 737-743 nm Amax
suitability
suitable for fluorescence
detection method
fluorometric
storage temp.
−20°C
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service