Skip to Content
Merck
All Photos(1)

Key Documents

798266

Sigma-Aldrich

Polydimethylsiloxane-diacrylamide

average Mn 1,200, contains MEHQ as inhibitor

Synonym(s):

PDMS crosslinker, PDMS dimethacrylamide

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
C16H32N2O3Si2(SiOC2H6)n
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

mol wt

average Mn 1,200

contains

MEHQ as inhibitor

refractive index

n20/D 1.433

transition temp

flash point >230 °C

density

0.998 g/mL

storage temp.

−20°C

Application

This macromonomer can be used in many applications, including applications where gas permeability is desired, such as ophthalmic applications. The methacrylamide functionality on both ends of the chain can react to form a crosslinked network. This methacrylamide can also be polymerized using controlled radical polymerization such as ATRP or RAFT.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service