808806
Tungsten disulfide
Crystal, 99.995%
Synonym(s):
Tungsten(IV) sulfide, Tungsten sulfide, Tungsten sulphide
Select a Size
Select a Size
About This Item
Recommended Products
Assay
99.995%
form
crystals
L × W × W
0.5 cm × 0.5 cm × 0.1-0.15 mm, typical size, some variation in dimensions may occur with different batches
density
7.5 g/mL at 25 °C (lit.)
SMILES string
S=[W]=S
InChI
1S/2S.W
InChI key
ITRNXVSDJBHYNJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Preparation Note
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Professors summarize recent 2D materials synthesis advancements and biosensing applications in various fields.
Advances in scalable synthesis and processing of two-dimensional materials
Research focuses on sustainable and cost-effective power generation systems to meet the growing demand for environmentally friendly energy sources.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service