Skip to Content
Merck
All Photos(1)

Key Documents

09166

SAFC

Mannitol Salt Agar Plates

suitable for microbiology

Synonym(s):

Chapman Agar Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Pricing and availability is not currently available.

shelf life

limited shelf life, expiry date on the label

composition

casein peptone, 5 g/L
D-mannitol, 10 g/L
meat extract, 1 g/L
meat peptone, 5 g/L
phenol red, 0.025 g/L
sodium chloride, 75 g/L

diam.

55 mm

final pH

7.4±0.2

application(s)

microbiology

storage temp.

2-8°C

Application

Prepared solid selective medium for the isolation of Staphylococci. 55 mm plates in blister packs. Each blister pack comes in a sealed flowpack bag. This kind of packaging avoids fast dehydration and contamination.

Packaging

package with 30 plates

Other Notes

Delivery time longer than 3 weeks.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service