93655
Tryptone Agar
suitable for microbiology, NutriSelect® Plus
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41171605
Recommended Products
form
powder
Quality Level
shelf life
limited shelf life, expiry date on the label
composition
agar, 15 g/L
sodium chloride, 8 g/L
tryptone, 10 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
application(s)
environmental
food and beverages
microbiology
suitability
nonselective for bacteria (General Media)
Application
Used for the determination of motility and fermentation ability of aerobic and anaerobic bacteria. Base for a wide variety of fastidious microorganisms.
Preparation Note
Dissolve 33 g in 1 litre distilled water. Autoclave at 121°C for 15 minutes.
Legal Information
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service