Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N1533

Sigma-Aldrich

Nitric Oxide Synthase, Endothelial bovine

recombinant, expressed in baculovirus infected Sf9 cells, buffered aqueous solution

Synonym(s):

NOS III, ecNOS

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

form

buffered aqueous solution

specific activity

≥0.3 units/mg protein

mol wt

135 (kDa/subunit, homodimer)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Biochem/physiol Actions

For measuring NOS activity in the presence of exogenously added co-factors and/or inhibitors.
NOS is responsible for the biosynthesis of nitric oxide from L-arginine and is constituitively expressed in endothelial cells (eNOS) and brain (nNOS).
Responsible for the biosynthesis of nitric oxide from L-arginine; constituitively expressed in endothelial cells (eNOS) and brain (nNOS).
Suitable for measuring NOS activity in the presence of exogenously added co-factors and/or inhibitors.

Unit Definition

One unit of enzyme produces 1 nmole of nitric oxide per minute at 37 °C in 50 mM HEPES, pH 7.4, containing 5 μM oxyhemoglobin, 1 mM CaCl2, 20 μg/ml calmodulin, 0.1 mM NADPH, 50 μM arginine, 12 μM tetrahydrobiopterin, and 170 μM DTT.

Physical form

Solution in 50 mM HEPES, pH 7.4, with 10% glycerol, 5 mM CHAPS, and 100 μM DTT.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-Ling Lai et al.
Journal of clinical biochemistry and nutrition, 46(2), 177-185 (2010-03-11)
N(omega)-Carboxymethyl-arginine (CMA), N(omega)-carboxyethyl-arginine (CEA) and N(delta)-(5-hydro-5-methyl-4-imidazolon-2-yl)-ornithine (MG-H1) have been identified as L-arginine-derived advanced glycation end products (AGEs) formed by non-enzymatic reactions between reducing sugars such as glucose and amino groups in proteins. These AGEs are structurally analogous to endogenous inhibitors
Silviya R Stateva et al.
PloS one, 10(4), e0120798-e0120798 (2015-04-02)
Calmodulin (CaM) phosphorylated at different serine/threonine and tyrosine residues is known to exert differential regulatory effects on a variety of CaM-binding enzymes as compared to non-phosphorylated CaM. In this report we describe the preparation and characterization of a series of
Ying-Ling Lai et al.
Journal of clinical biochemistry and nutrition, 50(1), 84-89 (2012-01-17)
The cholesterol ozonolysis products secosterol-A and its aldolization product secosterol-B were recently detected in human atherosclerotic tissues and brain specimens, and have been postulated to play pivotal roles in the pathogenesis of atherosclerosis and neurodegenerative diseases. We examined several oxidized
Heather S Bevan et al.
American journal of physiology. Renal physiology, 301(4), F733-F742 (2011-07-22)
Laminar shear stress is a key determinant of systemic vascular behavior, including through activation of endothelial nitric oxide synthase (eNOS), but little is known of its role in the glomerulus. We confirmed eNOS expression by glomerular endothelial cells (GEnC) in
Yunzhou Dong et al.
Journal of cellular and molecular medicine, 13(9A), 2899-2910 (2008-07-16)
Oxidation and glycation of low-density lipoprotein (LDL) promote vascular injury in diabetes; however, the mechanisms underlying this effect remain poorly defined. The present study was conducted to determine the effects of 'heavily oxidized' glycated LDL (HOG-LDL) on endothelial nitric oxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service