CMC0006
SIG10 ULTRA Electrocompetent Cells
for BAC & cosmid cloning
Sign Into View Organizational & Contract Pricing
All Photos(5)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.85
Recommended Products
biological source
Escherichia coli
grade
Molecular Biology
form
buffered aqueous solution
growth mode
adherent or suspension
morphology
rod shaped
technique(s)
microbiological culture: suitable
cell transformation
competent cell type: electrocompetent
transformation efficiency: ≥4 x 1010 cfu/μg
shipped in
dry ice
storage temp.
−70°C
General description
The SIG10 ULTRA Electrocompetent Cells have the highest transformation efficiency available. They are to be used for the most demanding cloning situations, such as construction of large, high complexity libraries or cloning difficult targets, which require the greatest number of transformants possible.
Genotype
F- mcrA Δ(mrr-hsdRMS-mcrBC) endA1 recA1 Φ80dlacZΔM15 ΔlacX74 araD139 Δ(ara,leu)7697galU galK rpsL nupG λ- tonA (StrR)
Genotype
F- mcrA Δ(mrr-hsdRMS-mcrBC) endA1 recA1 Φ80dlacZΔM15 ΔlacX74 araD139 Δ(ara,leu)7697galU galK rpsL nupG λ- tonA (StrR)
Features and Benefits
- share the most useful genetic elements of standard cloning strains like DH5α™ DH10B, JM109, TOP10, etc. and directly replace them in cloning protocols.
- provide the performance researchers need with ease of use.
- provide solutions for a wide range of applications at economical prices.
- Blue - white screening
Components
- SIG10 ULTRA electrocompetent cells
- pUC 19 transformation control DNA
- recovery medium for cloning
Legal Information
DH5α is a trademark of Invitrogen Corp.
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service