Skip to Content
Merck
All Photos(2)

Key Documents

S0189

Sigma-Aldrich

Stemline® Hematopoietic Stem Cell Expansion Medium

suitable for, Without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.81

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

suitability

suitable for

shipped in

wet ice

storage temp.

2-8°C

General description

Developed to promote the optimal expansion of human hematopoietic stem cells (HSC) from bone marrow, mobilized peripheral blood, and cord blood, Stemline Hematopoietic Stem Cell Expansion Medium demonstrates higher total nucleated cell (TNC) fold increases than other commercially available serum-free media formulations. Additionally, colony-forming unit (CFU) assays indicate that CD34+ hematopoietic stem cells grown in Stemline media demonstrate significantly greater expansion of bother early (HPP-CFC) and late (GM-CFC) progenitor populations when compared to those grown in alternative media. Human cord blood cells expanded in Stemline Media demonstrate impressive self-revewal when transplanted into immunodeficient NOC/SCID mice, illustrating Stemline′s utility in true functional trial.

Stemline Hematopoietic Stem Cell Expansion Medium free of serum and all other animal-derived components with the exception of human serum albumin and cholesterol. This exclusion increases performance consistency and elimates safety risks associated with potential adventitious agents.

Produced in a GMP state-of-the-art facility, Stemline Hematopoietic Stem Cell Expansion Medium is clearly an excellent choice for your HSC applications.

Application

Intended for the expansion of CD 34+ progenitors derived from bone marrow, mobilized peripheral blood and cord blood.

Reconstitution

Must be supplemented with growth factors and cytokines according to individual user protocols.

Legal Information

Stemline is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiming Chen et al.
British journal of haematology, 192(3), 568-576 (2020-12-21)
Multiple myeloma (MM) tumour cells evade host immunity through a variety of mechanisms, which may potentially include the programmed cell death ligand-1 (PD-L1):programmed cell death protein-1 (PD-1) axis. This interaction contributes to the immunosuppressive bone marrow (BM) microenvironment, ultimately leading

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service