Skip to Content
Merck
All Photos(1)

Key Documents

SHC009

Sigma-Aldrich

MISSION® pLKO.1-puro ARHGDIA shRNA Control Plasmid DNA

Positive control targeting ARHGDIA

Synonym(s):

MISSION® Control Vectors

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

MDL number:
UNSPSC Code:
12352200

product line

MISSION®

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This control is intended for use as a positive control for knockdown of human gene targets when used with clones of the MISSION TRC human shRNA collections. The DNA format of this control construct permits direct transfection in knockdown experiments or use in the viral packaging process (for lentiviral-based knockdown experiments).

The MISSION Rho GDP Dissociation Inhibitor (GDI) alpha (ARHGDIA) shRNA Control Vector is a lentiviral-based plasmid vector that contains a short hairpin sequence targeting the human Rho GDP Dissociation Inhibitor (GDI) alpha transcript (Gene Symbol: ARHGDIA). Rho GDP dissociation inhibitor (GDI) alpha (ARHGDIA) is a ubiquitously expressed protein that acts on human Rho GTPases including RhoA, Rac1 and Cdc42, by keeping these proteins locked in an inactive state. Complete understanding of ARHGDIA′s roles is still being elucidated but it is believed to be involved in various signal transduction pathways and cellular cytoskeleton functions. The vector backbone of the ARHGDIA clone is the same as the MISSION TRC shRNA collections (pLKO.1). The specific clone number for the ARHGDIA control is TRNC0000008004 and was chosen from the available complete target set (NM_004309). The control has demonstrated consistent knockdown of 90% compared to normal gene expression in the human epithelial lung carcinoma cell line A549 via quantitative RT PCR and Western blot analysis (Antibody Product No. R3025).

Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells respectively. In addition, self-inactivating replication incompetent viral particles can be produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids (Product No. SHP001). The MISSION Rho GDP Dissociation Inhibitor (GDI) alpha (ARHGDIA) shRNA Control Vector is provided as 10 μg of plasmid DNA in Tris-EDTA (TE) buffer at a concentration of 500 ng/μl.

Application

To see more application data, protocols, vector maps visit sigma.com/shrna.

Legal Information

Use of this product is subject to one or more license agreements. For details, please see http://sigmaaldrich.com/missionlicense.
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruth S Cruz Cosme et al.
Journal of virology, 83(7), 2839-2850 (2009-01-16)
Human cytomegalovirus (HCMV), a member of the beta subgroup of the family Herpesviridae, causes serious health problems worldwide. HCMV gene expression in host cells is a well-defined sequential process: immediate-early (IE) gene expression, early-gene expression, DNA replication, and late-gene expression.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service