2879901-U
Inlet Liner, Split/Splitless Type, Single Taper FocusLiner™ Design (wool packed)
pkg of 1 ea
About This Item
Recommended Products
packaging
pkg of 1 ea
L × O.D. × I.D.
78.5 mm × 6.3 mm × 4.0 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Fits Agilent (5890, 6890, and 7890)
- Fits Finnigan (9001GCQ)
- Fits Varian (CP-1177 Injector)
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Pistachios 45% fat complicates pesticide analysis. But Z-Sep+ sorbent cleanup after QuEChERS extraction can reduce background for reliable results.
Dual-layer cartridge achieves efficient SPE clean-up for challenging pesticide residue testing in turmeric and spices.
2-layer SPE cartridge efficiently reduces background for ginger contaminant analysis.
Assessment of clean-up sorbents for extracting PAHs from salmon and PCBs from milk, optimized for GC analysis with MS and ECD detection.
Protocols
Paprika spice's PAH contamination risk due to environmental exposure highlights food safety concerns.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service