MilliporeSigma
HomeSolid Phase Extraction (SPE)Pesticide Analysis in Turmeric by LC/MS/MS & GC/MS/MS
Pesticide Analysis in Turmeric by LC/MS/MS & GC/MS/MS
Katherine K. Stenerson,  Principal R&D Scientist
Introduction
Turmeric is a plant indigenous to south Asia, with a majority of its production coming from India. The rhizome of the plant is used to produce powdered turmeric, which is used in foods, cosmetics, and some medicines. It is also an essential constituent of curry, which is a mixture of spices used extensively in Indian cooking. Turmeric has also been used in traditional medicines for thousands of years and recently has garnered attention for studies showing its potential antioxidant, anti-inflammatory, antimutagenic, antimicrobial, and anticancer properties.
Pesticide residue testing of turmeric and other spices is required by many countries. For example, Canada has set maximum residue limits for 42 different pesticides in turmeric root. The U.S. EPA has set tolerance limits for a variety of pesticides in root and tuberous vegetables, of which turmeric is included.
Turmeric contains more than 100 different components, with two of the main constituents being curcumin and volatile oils. Curcumin gives turmeric its distinctive yellow/orange color, while the volatile oils consist primarily of terpenes. Turmeric also contains some fats; specifically, sterols and fatty acids. This complex composition makes extracts produced from turmeric a challenge in the chromatographic analysis of pesticides, as residual pigments and oils can contaminate both GC/MS and LC/MS systems.
When dealing with very high background samples such as turmeric, standard QuEChERS cleanup may not offer enough capacity. For better cleanup, solid phase extraction (SPE), including dual-layer cartridges, can be used. These cartridges often contain graphitized carbon black (GCB) in the top bed and primary-secondary amine (PSA) in the bottom bed. PSA retains acidic interferences such as fatty acids. GCB removes planar molecules such as pigments and sterols. Common GCBs, however, will retain all molecules with planar structures, including some pesticide analytes such as hexachlorobenzene. To increase recoveries of these pesticides, toluene is normally added to the elution solvent. However, there are issues associated with the use of toluene. It can affect the ability of the PSA to retain fatty acids, and its presence in the final extract is problematic for HPLC analysis.
In this application, a different dual-layer SPE cartridge was used in the cleanup of extracts of turmeric powder prior to pesticide analysis by GC/MS/MS and LC/MS/MS. This cartridge, the Supelclean™ Ultra 2400, was designed for the cleanup of acetonitrile extracts made from difficult matrices such as dry commodities (spices, tea, etc.) prior to pesticide residue analysis. The top bed consists of a mixture of PSA, C18 and a graphitized, spherical carbon known as Graphsphere™ 2031. This carbon was engineered to remove sufficient pigmentation while allowing for better recoveries of planar compounds, without the need for toluene in the elution solvent. The bottom layer of the cartridge contains Z-Sep, a zirconia-coated silica. Z-Sep removes oily residues and provides additional retention of some pigments. The combination of these sorbents in an SPE format offers more capacity than QuEChERS cleanup, and compared to traditional GCB/PSA dual layer cartridges, does not require the use of toluene in the elution solvent to recover planar pesticides.
Experimental
Turmeric powder was obtained from a local grocery store. Samples were spiked at 100 ng/g with the pesticides listed in Tables 1 and 2. Sample extracts were prepared and cleaned following the procedures in Figure 1. A set of 3 spiked samples and 1 unspiked (blank) were prepared and analyzed for each set of pesticides. Analysis was done by GC/MS/MS and LC/MS/MS using the conditions listed in Tables 3 and 4 (with MS/MS transitions shown in Tables 1 and 2). Quantitation was performed against multi-point calibration curves prepared in unspiked turmeric extract (after cleanup). Recoveries were calculated as the average of the three spiked replicates, less anything found in the unspiked extract. No internal standards were used, thus the values reported represent absolute recoveries.
Results and Discussion
Background
Prior to cleanup, the extract appeared orange-brown in color with a yellow oily residue (Figure 2). After cleanup for both LC and GC, the extracts appeared substantially lighter and clearer. Figures 3 and 4 show a comparison between extracts at the same level of dilution with and without cleanup. The LC extract (in 80% aqueous) was almost devoid of color, with very little cloudiness. The extract for GC analysis was a pale yellow color, with substantially less oily residue. Full scan GC/MS analyses of GC extracts are shown in Figure 5 as total ion chromatograms (TICs). The peak pattern is similar between the two, with the main peaks consisting primarily of terpenes. These compounds are easily volatilized in the GC inlet, and do not pose issues with system contamination, however they can interfere with mass spectral detection, requiring the use of MS/MS for selectivity. The overall amplitude of the peaks was less after cleanup, as is shown by a 21% reduction in the peak area sums for each in Figure 5.
 MRM 1CEMRM 2CE
Alachlor
188/16010188/13040
Aldrin263/19335263/19135
γ-BHC183/14715181/1455
Azinphos-methyl160/7715132/7715
Chloropyrifos314/2865314/25815
Chloropyrifos-methyl286/9320288/9320
Cypermthrins165/9110163/9110
4,4'-DDT235/19915235/16525
Diazinon199/13515137/8410
Dichlorvos185/9325145/10925
Dimethoate125/792093/6310
Disulfoton88/60588/59
15
Endosulfan β241/20615
241/17030
Endosulfan-α241/20615241/17030
Ethion231/129
20121/6510
Fenitrothion277/12520277/10920
Heptachlor274/23915272/23715
Hexachlorobenzene284/24920284/21435
Iprodione I314/5635187/12425
Iprodione II316/5635187/12425
Malathion173/9915158/1255
Metalaxyl234/17410234/14620
Methoxychlor227/16930227/141
30
Mevinphos192/12725192/10925
Parathion-methyl233/10910124/4710
Permethrins183/16810183/16510
Phenthoate274/12515274/12110
Phorate260/755231/12925
Phosalone182/10215182/7530
Pirimiphos-methyl290/15120290/12525
Profenophos339/26915339/18815
Quintozene295/23720237/14330
Vinclozolin212/14530187/12420
Table 1 Pesticides Studied in Turmeric Powder by GC/MS/MS Analysis
 MRM  
Frag (V)CE (V)Cell Acc (V)
Acephate184/1437005
Acetamiprid223.1/12680272
Boscalid (Nicobifen)343/307.1145166
Carbendazim (Azole)192.1/160.1105162
Chlorbufam224/172.0212053
Cycluron199.2/72120202
Diflubenzuron311/1588082
Fenoxanil329.08/18980303
Fosthiazate284/6190602
Methabenzthiazuron
222.1/165.190122
Methamidophos142/12585102
Methomyl163.1/1065042
Monocrotophos (Azodrin)224.1/19365
0
5
Nitralin346.11/304100103
Oxamyl237.1/7260122
Pirimicarb239.15/72.1100202
Procymidon301/284*7082
Propaquizafop444.12/100.1125162
Tetraconazole372/159130362
Uniconazole-P292.1/125135402
Table 2Pesticides Studied in Turmeric Powder by LC/MS/MS Analysis * Ammonium adduct
column:SLB®-5ms, 30 m × 0.25 mm ID, 0.25 μm (28471-U)
oven:50 °C (2 min), 8 °C/min to 320 °C (5 min)
inj. temp.:250 °C
carrier gas:helium, 1.4 mL/min, constant
detector:MRM (table 3)
MSD interface:320 °C
injection :1μL, splitless (splitter open at 0.75 min)
liner:4 mm I.D., split/splitless type, single taper wool packed FocusLiner™ design (2879901-U)
Table 3GC/MS/MS Run Conditions for the Analysis of Pesticides in Turmeric
column:Ascentis® Express C18, 10 cm × 2.1 mm ID, 2 μm
mobile phase:[A] 5 mM ammonium formate, 0.1% formic acid in water;
[B] 5 mM ammonium formate, 0.1% formic acid in methanol
gradient:95% A, 5% B held for 1 min; to 50% A in 3 min; to 100%
B in 8 min; held for 1 min;, to 95% A in 1.5 min; held at 95%
A for 1.5 min
flow rate:0.4 mL/min
detector:MRM (table 2)
injection:5 μL
Table 4 LC/MS/MS Run Conditions for the Analysis of Pesticides in Turmeric
Extraction and cleanup procedure used for turmeric powder
Figure 1.Extraction and Cleanup Procedure Used for Turmeric Powder, GC and LC
Undiluted Acetonitrile Extract
Figure 2. Undiluted Acetonitrile Extract of Turmeric Powder Before Cleanup
Turmeric Extracts
Figure 3. Turmeric Extracts at the Same Dilution (167X total); Without Cleanup, and After Cleanup for LC/MS/MS Analysis    
Turmeric Extracts at the Same Dilution
Figure 4. Turmeric Extracts at the Same Dilution (5X); Without Cleanup, and After Cleanup for GC/MS/MS Analysis
Pesticide Recovery and Reproducibility
The average recoveries obtained from spiked turmeric samples (n=3) are presented in Table 5. Of the 51 pesticides spiked, all except hexachlorobenzene had recovery of greater than 70%. Hexachlorobenzene, a pesticide with a planar structure, was recovered at 67% after cleanup. It should be noted that this was without using toluene in the elution solvent, as is necessary to obtain good recoveries from dual-layer cartridges containing graphitized carbon black. Although not shown here, higher recovery of hexachlorobenzene has been obtained by loading more turmeric extract (300 μL) on the Supelclean™ Ultra 2400 cartridge. This indicates that the presence of more matrix displaced the hexachlorobenzene, thus reducing its retention on the carbon. However the higher sample loading produced an extract with more color, a sign that the cleanup capacity of the cartridge had been reached or exceeded for this matrix.
PesticideAvg. RecoveryRSDAnalysis
Alachlor
99%23%GC/MS/MS
Aldrin85%10%GC/MS/MS
Azinphos-methyl89%11%GC/MS/MS
γ-BHC83%8%GC/MS/MS
Chloropyrifos96%12%GC/MS/MS
Chloropyrifos-Methyl113%6%GC/MS/MS
Cypermthrin (isomer 1)99%15%GC/MS/MS
4,4'-DDT95%8%GC/MS/MS
Diazinon92%14%GC/MS/MS
Dichlorvos78%31%GC/MS/MS
DIsulfoton86%7%GC/MS/MS
Endosulfan β86%35%GC/MS/MS
Endosulfan-α92%23%GC/MS/MS
Ethion97%7%GC/MS/MS
Fenitrothion63%5%GC/MS/MS
Heptachlor81%7%GC/MS/MS
Hexachlorobenzene67%9%GC/MS/MS
Iprodione (isomer 1)103%5%GC/MS/MS
Malathion90%10%GC/MS/MS
Metalaxyl86%21%GC/MS/MS
Methoxychlor78%12%GC/MS/MS
Mevinphos73%7%GC/MS/MS
Parathion-Methyl88%8%GC/MS/MS
Permethrin (isomer 1)104%24%GC/MS/MS
Phenthoate89%7%GC/MS/MS
Phorate82%10%GC/MS/MS
Phosalone
90%
7%GC/MS/MS
Pirimiphos-methyl74%3%GC/MS/MS
Profenophos88%7%GC/MS/MS
Quintozene75%8%GC/MS/MS
Vinclozolin90%6%GC/MS/MS
Acephate89%6%LC/MS/MS
Acetamiprid102%4%LC/MS/MS
Boscalid (Nicobifen)86%7%LC/MS/MS
Carbendazim (Azole)106%7%LC/MS/MS
Chlorbufam92%18%LC/MS/MS
Cycluron103%5%LC/MS/MS
Diflubenzuron101%5%LC/MS/MS
Fenoxanil91%10%LC/MS/MS
Fosthiazate95%4%LC/MS/MS
Methabenzthiazuron96%4%LC/MS/MS
Methamidophos85%5%LC/MS/MS
Methomyl106%
6%LC/MS/MS
Monocrotophos (Azodrin)97%3%LC/MS/MS
Nitralin124%55%LC/MS/MS
Oxamyl104%3%LC/MS/MS
Pirimicarb97%3%LC/MS/MS
Procymidon
91%13%LC/MS/MS
Propaquizafop97%1%LC/MS/MS
Tetraconazole98%2%LC/MS/MS
Uniconazole-P103%19%LC/MS/MS
Table 5 Pesticide Recoveries and % RSD Values (n=3) for Spiked Replicates; Turmeric Spiked at 100 ng/g
GC/MS Scan Analyses of Turmeric Extracts
Figure 5.GC/MS Scan Analyses of Turmeric Extracts Before and After Cleanup with Supelclean™ Ultra 2400 Shown with same Y-scale. Sum of area counts for all peaks is indicated with each.
Recoveries Within Indicated Percent Relative
Number of Pesticides with Average Recoveries
Figure 6. Number of Pesticides with Average Standard Deviation (%RSD) Ranges After Cleanup with Supelclean™ Ultra 2400 (Recoveries from turmeric powder spiked at 100 ng/g.)
Conclusion
A new cleanup method has been developed using the Supelclean™ Ultra 2400 dual-layer SPE cartridge. The selection of sorbents in this cartridge allows for cleanup of acetonitrile extracts of very difficult samples such as spices and other dry commodities. The Graphsphere™ 2031 carbon used in the upper layer removes/reduces pigmentation while still allowing for recovery of planar pesticides without the use of toluene in the elution solvent. Z-Sep sorbent in the bottom layer of the cartridge removes oils and some pigments, as was indicated in the cleanup of turmeric extracts for both GC and HPLC analysis. Suitable recoveries for a wide range of pesticides of different polarities and classes were obtained from turmeric extract, and minimal background interference was noted. In this work, a 1 mL Supelclean™ Ultra 2400 cartridge was used. A larger 3 mL version of the cartridge is also available which can accommodate a higher sample loading.
Acknowledgements
The author would like to thank Richard Schriner and Bruce Morris of R.J. Hill Laboratories for their helpful discussions and input.
Materials
Loading
References
1.
Prasad S, Aggarwal B. 2011. Turmeric, the Golden Spice.263-288. https://doi.org/10.1201/b10787-14
2.
2016. Health Canada MRL database. [Internet]. Available from: http://www.hc-sc.gc.ca
3.
2016. Electronic Code of Federal Regulations (eCFR) Title 40, Chapter 1, Subchapter E, Part 180. [Internet]. Available from: https://www.ecfr.gov/cgi-bin/text-idx?tpl=/ecfrbrowse/Title40/40cfr180_main_02.tpl
4.
2011. Chemical composition and product quality control of turmeric (Curcuma longa L.). [Internet]. Available from: https://core.ac.uk/download/pdf/72733173.pdf
5.
Shimelis O, Yang Y, Stenerson K, Kaneko T, Ye M. 2007. Evaluation of a solid-phase extraction dual-layer carbon/primary secondary amine for clean-up of fatty acid matrix components from food extracts in multiresidue pesticide analysis. Journal of Chromatography A. 1165(1-2):18-25. https://doi.org/10.1016/j.chroma.2007.07.037