56059
7-Hydroxy-N-octadecylcoumarin-3-carboxamide
suitable for fluorescence, ≥98.0% (TLC)
About This Item
Recommended Products
Assay
≥98.0% (TLC)
solubility
chloroform/DMSO (1:1): soluble
fluorescence
λex 359 nm; λem 406 nm in chloroform/DMSO (1:1)
suitability
suitable for fluorescence
SMILES string
CCCCCCCCCCCCCCCCCCNC(=O)C1=Cc2ccc(O)cc2OC1=O
InChI
1S/C28H43NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20-29-27(31)25-21-23-18-19-24(30)22-26(23)33-28(25)32/h18-19,21-22,30H,2-17,20H2,1H3,(H,29,31)
InChI key
CVZKNSNXXVFUMZ-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service