Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV39934

Sigma-Aldrich

Anti-ZNF555 (AB4) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC26707, Anti-Zinc finger protein 555

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

73 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF555(148254)

Immunogen

Synthetic peptide directed towards the C terminal region of human ZNF555

Biochem/physiol Actions

ZNF555 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 15 C2H2-type zinc fingers and 1 KRAB domain. ZNF555 may be involved in transcriptional regulation.

Sequence

Synthetic peptide located within the following region: LRRHVRIHTTEKQYKCNVGHPPANEFMCSASEKSHQERDLIKVVNMVLPL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service