Skip to Content
Merck
All Photos(1)

Key Documents

PHL83278

Columbianadin

phyproof® Reference Substance

Synonym(s):

Columbianedin, Columbianetin angelate, Zosimin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20O5
CAS Number:
Molecular Weight:
328.36
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

Pricing and availability is not currently available.

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

storage temp.

2-8°C

SMILES string

O=C1OC2=C(C=CC3=C2C[C@@H](C(C)(C)OC(/C(C)=C\C)=O)O3)C=C1

InChI

1S/C19H20O5/c1-5-11(2)18(21)24-19(3,4)15-10-13-14(22-15)8-6-12-7-9-16(20)23-17(12)13/h5-9,15H,10H2,1-4H3/b11-5-/t15-/m0/s1

InChI key

JRIBPWOXWIRQOQ-GHAIFCDISA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: angelica

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service