Skip to Content
Merck
All Photos(2)

Key Documents

E7158

Sigma-Aldrich

pFLAG-CMV-4 Expression Vector

shuttle vector for intracellular transient or stable expression of N-terminal Met-FLAG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Pricing and availability is not currently available.

tag

Met-FLAG tagged

grade

Molecular Biology

form

buffered aqueous solution

Peptide tag location

N-terminal

shipped in

dry ice

storage temp.

−20°C

General description

pFLAG-CMV-4 is a 6.2 kb expression vector used for transient or stable expression in mammalian cells. The vector is a derivative of pCMV5 used for transient or stable expression of a properly inserted open reading frame as an N-terminal Met-FLAG® fusion protein.

The pFLAG-CMV-4 Expression Vector is a shuttle vector containing both bacterial and SV40 origins of replication for propagation in both E.coli and mammalian cells. Efficiency of replication and genomic integration is optimal when using host cells that express the SV40 large T antigen.

The FLAG epitope is a small, hydrophilic 8 amino acid tag (DYKDDDDK) that provides for sensitive detection and high quality purification using ANTI-FLAG products available for Sigma Aldrich.

pFLAG-CMV-4-BAP Control Plasmid is a 7.7 kb derivative of pCMV5 used for transient intracellular expression of N-terminal Met-FLAG bacterial alkaline phosphatase fusion protein in mammalian cells.

Vector Maps and Sequences

Components

  • pFLAG-CMV-4 Expression Vector 20 μg (E1775) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
  • pFLAG-CMV-4-BAP Control Plasmid 20 μg (C4722) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Principle

The promoter-regulatory region of the human cytomegalovirus2 drives transcription of FLAG®-fusion constructs. The aminoglycoside phosphotransferase II gene (neo-r) confers resistance to aminoglycosides such as G 418 allowing for selection of stable transfectants.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Palgrave et al.
Journal of virology, 85(12), 6008-6014 (2011-04-01)
African swine fever virus (ASFV) is a highly infectious disease of domestic pigs, with virulent isolates causing a rapidly fatal hemorrhagic fever. In contrast, the porcine species endogenous to Africa tolerate infection. The ability of the virus to persist in
Tadashi Asakura et al.
International journal of oncology, 33(2), 389-395 (2008-07-19)
We investigated the induction of apoptosis via deamidation of Bcl-xL and translocation of Bax to the mitochondria by treatment with GSH-DXR. GSH-DXR treatment of HepG2 cells, which did not express GST P1-1, exhibited deamidation of Bcl-xL, and the degree of
Chunxue Lu et al.
PloS one, 8(7), e68764-e68764 (2013-07-31)
Glycogen has been localized both inside and outside Chlamydia trachomatis organisms. We now report that C. trachomatis glycogen synthase (GlgA) was detected in both chlamydial organism-associated and -free forms. The organism-free GlgA molecules were localized both in the lumen of
M G Chaitra et al.
Journal of medical microbiology, 56(Pt 4), 466-474 (2007-03-22)
The PE and PPE proteins of Mycobacterium tuberculosis form a source of antigenic variation among different strains of M. tuberculosis. One of the PE_PGRS proteins, Rv1818c, plays a role in the pathogenesis of mycobacterial infection and specifically influences host-cell responses
Junya Masumoto et al.
The Journal of biological chemistry, 278(6), 4268-4276 (2002-12-05)
The pyrin domain was identified recently in multiple proteins that are associated with apoptosis and/or inflammation, but the physiological and molecular function of these proteins remain poorly understood. We have identified Caspy and Caspy2, two zebrafish caspases containing N-terminal pyrin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service