Skip to Content
Merck
All Photos(1)

Key Documents

ECM221

Sigma-Aldrich

QCM Laminin Migration Assay (24-well, fluorometric)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.76

technique(s)

cell based assay: suitable

Quality Level

detection method

fluorometric

Application

The Millipore QCM Laminin Migration Assay incorporates a 5 µm porous membrane Boyden chamber and mouse laminin, a major extracellular matrix (ECM) protein in basement membrane, to examine cell migration dependent upon the presence of laminin. The laminin is isolated from mouse sarcoma and it is enriched with mouse laminin 1 (laminin-111). This ECM promotes cell adhesion, proliferation as well as migration in many cell types such as embryonic stem cells, neural cells, liver cells and cancer cells. Cells interact with laminin through integrins α1β1, α2β2, α3β1, α6β1, α7β1, and α6β4. The assay is designed specifically to examine the migration of a subset of fibroblasts, cancer cells and neural cells.
Laminin protein is coated on the bottom of each chamber′s porous membrane as the driver for cell migration. A similarly BSA-coated control chamber(s) provides an appropriate migration control. Cells that migrate toward laminin are stained using Calcein-AM a living cell fluorescent dye, quantified on a fluorescent microplate reader.

Each kit provides sufficient materials for the evaluation of 24 samples.

Packaging

Sufficient for 12 assays

Components

ECM221-1 1. 24-well Cell Migration Plate Assembly - 5 µm: (Part No. 2005708) Two 24-well plates, each containing 12- 5 µm pore inserts per plate. 2. BSA 30% solution: (Part No. CS203352) One vial containing 0.5 mL of a 30% solution. 3. 24-well Stain Receiver Plate: (Part No. CR209941) One each. 4. 96-Well Black Quantitation Plate: (Part No. CR201763) One each. 5. Forceps: 1 pair ECM221-2 1. Laminin: (Part No. CS202685) Two vials each containing 60 μg in solution. 2. Accutase®: (Cat. No. SCR005) One 100 mL bottle of cell detachment solution. 3. Calcein-AM: (Part No. CS202541) One 50 µg vial.

Storage and Stability

ECM221 should be used within four months from date of receipt. In that time:• ECM221-1 kit components should be stored at 2° to 8°C. DO NOT FREEZE.• ECM221-2 kit components must be stored at -20°C until needed.

Legal Information

Accutase is a registered trademark of Innovative Cell Technologies, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service