50448
Staphylo Monotec test kit Plus
suitable for microbiology
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41171621
NACRES:
NA.85
Recommended Products
technique(s)
microbe id | immunological: suitable
microbe id | specific enzyme detection: suitable
application(s)
clinical testing
food and beverages
microbiology
storage temp.
2-8°C
suitability
Staphylococcus spp.
Application
Staphylo Monotec test kit Plus is a new rapid agglutination test for differentiation between Staphylococcus aureus and other Staphylococcus. This method has been evaluated for clinical specimes and food material. With this kit coagulase and protein A and capsular Polysaccharide (serotype 5) on S. aureus can be detect in one step. Staphylo Monotec test kit Plus has an increased sensitivity and specificity compared to the old Staphylo Monotec test kit which results in detecting more methicillin resistant Staphyloccocus aureus.
Components
1 x Test reagent (Cat.No. 88786; vial with red cap), 1.9 ml
1 x Control reagent (Cat. No. 69187; vial with white cap), 1.9 ml
25 x Analysis cards
(contains reagent for more than 100 tests)
1 x Control reagent (Cat. No. 69187; vial with white cap), 1.9 ml
25 x Analysis cards
(contains reagent for more than 100 tests)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service