S2022
Daminozide
suitable for plant cell culture
Synonym(s):
Succinic acid mono(2,2-dimethylhydrazide)
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
Linear Formula:
HO2CCH2CH2CONHN(CH3)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
1863230
EC Number:
MDL number:
UNSPSC Code:
12352200
Recommended Products
technique(s)
cell culture | plant: suitable
mp
162-164 °C (lit.)
storage temp.
2-8°C
SMILES string
CN(C)NC(=O)CCC(O)=O
InChI
1S/C6H12N2O3/c1-8(2)7-5(9)3-4-6(10)11/h3-4H2,1-2H3,(H,7,9)(H,10,11)
InChI key
NOQGZXFMHARMLW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service