Skip to Content
Merck
All Photos(1)

Key Documents

94327

Sigma-Aldrich

Deoxo-Fluor solution

50% in THF

Synonym(s):

Bis(2-methoxyethyl)aminosulfur trifluoride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14F3NO2S
CAS Number:
Molecular Weight:
221.24
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

form

liquid

concentration

50% in THF

density

1.03-1.05 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

COCCN(CCOC)S(F)(F)F

InChI

1S/C6H14F3NO2S/c1-11-5-3-10(4-6-12-2)13(7,8)9/h3-6H2,1-2H3

InChI key

APOYTRAZFJURPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Fluorination reactions
  • Preparation of fluorinated catechols via Umpolung based nucleophilic deoxyfluorination
  • Preparation of imidazole derivatives as CXCR3 antagonists
  • Ring expansion reactions
  • Synthesis of acyl azides from carboxylic acids
  • Aldehydes or ketones from carboxylic acids and Grignard reagents via in situ generation of Weinreb amides

Other Notes

Fluorinating agent with high thermal stability

Legal Information

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

<50.0 °F - closed cup

Flash Point(C)

< 10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fluorinating agent with high thermal stability.
S. Lal-Gauri et al.
The Journal of Organic Chemistry, 64, 7048-7048 (1999)
R.P. Singh, J.M. Shreeve
Journal of Fluorine Chemistry, 116, 23-23 (2002)
Lal et al.
The Journal of organic chemistry, 65(16), 4830-4832 (2000-08-24)
A variety of thiocarbonyl derivatives (thioketone, thioester, thioamide, dithioester, and dithiocarbamate) were converted to the corresponding gem-difluorides in excellent yields on reaction with the fluorinating agent, bis(2-methoxyethyl)aminosulfur trifluoride, in the presence of SbCl(3).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service