Skip to Content
Merck
All Photos(1)

Key Documents

6.17852

Supelco

Paraffin wax

with ceresin congealing point 60°C

Synonym(s):

Paraffin wax

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12181503

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Silicone oils aided fabrication of paraffin wax coated super-hydrophobic sand: A spectroscopic study.: This study employs silicone oils in the production of paraffin wax-coated super-hydrophobic sand, analyzed through spectroscopic techniques, suggesting applications in environmental and construction industries (Al-Mokhalelati et al., 2023).

  • Paraffin-Multilayer Graphene Composite for Thermal Management in Electronics.: This article explores the use of a paraffin-multilayer graphene composite for enhancing thermal management in electronic devices, contributing to improved performance and longevity of electronic components (Balan et al., 2023).

  • Spectroscopic investigations to reveal synergy between polystyrene waste and paraffin wax in super-hydrophobic sand.: The study investigates the synergistic effects of polystyrene waste and paraffin wax in creating super-hydrophobic sand, utilizing spectroscopic methods to uncover the interactions and potential for environmental applications (Al-Mokhalelati et al., 2023).


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service