Purity
98%
Country of Origin
CN
Inchi Code
1S/C9H20/c1-6-8(2)7-9(3,4)5/h8H,6-7H2,1-5H3
Inchi Key
AFTPEBDOGXRMNQ-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under 2,2,4-Trimethylhexane
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.