MilliporeSigma
Plasmid DNA | Plasmid Purification Kits
Circular plasmid DNA strand, showcasing the double helix structure with alternating colors representing the base pairs.
Plasmid purification is commonly used for molecular cloning, transformation, and recombinant protein expression. Our plasmid purification kit essentials are available based on the size of the bacterial culture and corresponding plasmid yield (miniprep, midiprep, maxiprep, megaprep, and gigaprep). Our plasmid kits provide reliable isolation of high-quality plasmid DNA. These kits are designed with advanced silica bind and elute technology. Additionally, our kits are compatible with both vacuum and spin formats and formulated without phenol/chloroform.
For incremental innovations, day after day, choose plasmid DNA purification kits and essentials that are the foundation of great work, not a variable that undermines it. Our plasmid purification kits allow researchers to maximize the reproducibility of their workflow and minimize the time spent obtaining plasmid DNA for transfection, sequencing, PCR, and other downstream applications. Explore all of our plasmid DNA purification kits below.
Products
Compare
Product Number
Product Name
Product Description
Pricing
GenElute™ plasmid miniprep kits
GenElute™ plasmid miniprep kits offer simple, rapid, and cost-effective methods for isolating plasmid DNA from E. coli cultures. These kits combine silica-based membrane technology and the convenience of a spin column format.
Features:
  • 40% more preps per kit than the leading supplier
  • Purify up to 20 µg of plasmid DNA per mL of culture
  • Purified plasmid DNA in less than 30 minutes for up to 24 preps
  • No phenol/chloroform extraction or alcohol precipitation required
GenElute™ HP plasmid prep kits
GenElute™ HP plasmid purification kits yield high-quality plasmid DNA in less than 30 minutes for Mini, Midi, and Maxiprep kits, or 1.5 hours for Mega and Gigaprep kits.
  • From harvested bacterial culture to pure plasmid DNA in less time
  • Up to 25 µg (Mini), 350 µg (Midi), 1.2 mg (Maxi), 5 mg (Mega), and 15 mg (Giga) yield of high-copy plasmid DNA
  • Offers the flexibility of a vacuum or spin format (Mega and Gigaprep in vacuum format only)
  • No phenol/chloroform extraction or alcohol precipitation required
  • Kits are stable at room temperature for convenient storage
GENOPURE™ PLASMID PREP KITS
Roche Genopure™ midi and maxi prep kits use anion exchange chromatography for plasmid purification. A modified alkaline lysis protocol optimized for cell cultures grown in LB media is used to isolate DNA. The lysate can be filtered rather than centrifuged prior to anion exchange, resulting in shorter plasmid prep times and complete removal of SDS from the purified product.
  • Reduce DNA shearing by clarifying lysate using pre-folded filters rather than centrifugation prior to anion exchange.
  • Save time with ready-to-use reagents (10 minutes hands-on time/75 minutes overall).
  • Purify all sizes and types of plasmid, even BAC DNA.
  • Process multiple samples in parallel using high-speed gravity-flow columns.
  • Eliminate the use of hazardous organic compounds such as cesium chloride, phenol, chloroform, and ethidium bromide.
GenElute™ endotoxin-free plasmid prep kits
Endotoxins reduce cell viability and transfection efficiencies among different eukaryotic cell lines and can influence the outcome and reproducibility of a given transfection experiment when using mammalian and other eukaryotic cells. GenElute™ endotoxin-free plasmid prep kits offer a simple, rapid, cost-effective method for purifying plasmid DNA with <0.1
EU/mg DNA for high-efficiency transfection.
  • Higher transfection efficiency and yield than market leaders
  • Purify endotoxin-free plasmid DNA (≤0.1 EU/mg DNA)
GenElute™ five-minute plasmid miniprep kits
GenElute™ five-minute miniprep kits feature a streamlined protocol yielding up to 5 mg high-quality plasmid DNA in about five minutes.
  • Only 5 minutes from start to finish
  • No pelleting cells or clearing lysates
  • Binding column works with any standard laboratory vacuum manifold
  • Choose vacuum or spin protocols
Related Resources
Page 1 of 3
Web Tools