698830
Graphite
platelet nanofibers, D × L 50-250 nm × 0.5-5 μm, 98% carbon basis
Select a Size
Select a Size
About This Item
Recommended Products
Assay
98% carbon basis
form
powder
composition
carbon, >99% trace metals analysis
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
D × L
50-250 nm × 0.5-5 μm
particle size
100 nm (average width, TEM)
2.5 μm (average length, TEM)
mp
3652-3697 °C (lit.)
greener alternative category
SMILES string
[C]
InChI
1S/C
Looking for similar products? Visit Product Comparison Guide
General description
Application
Preparation Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service