Skip to Content
Merck
All Photos(1)

Key Documents

40561

Millipore

β-Lactamase Testkit

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621

Pricing and availability is not currently available.

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages
microbiology

storage temp.

2-8°C

General description

β-Lactamase Testkit is a rapid acidimetric test for detection of β-lactamase activity of microorganisms. The test is performed in microtitre wells. This test is based on hydrolysis of the β-lactam ring in benzylpenicillin, which results in the production of penicilloic acid. This process causes acidification of the bacterial suspension, and changes the color of the acid-based indicator. The acidimetric test for beta-lactamase activity is suitable only for the detection of Haemophilus influenzae, Neisseria gonorrhoeae and Staphylococcus spp. This test is not suitable for the detection of the β-lactamase activity of other microorganisms (Branhamella catarrhalis, Enteroccocus faecalis, Neisseria meningitis, Enterococcus spp. and others). The result of the reaction is read after 10-30 min.

Components

Kit contains two microtitre plate strips coated with benzylpenicilline, diluent 75 mL and instructions sheet (for 192 tests)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service